missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCR V beta 3.1 Monoclonal Antibody (8F10), FITC
GREENER_CHOICE

Catalog No. ENTCR2740
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENTCR2740 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENTCR2740 Supplier Invitrogen™ Supplier No. TCR2740
Only null left

Mouse Monoclonal Antibody

TCR2740 targets TCR V beta 3.1 in FACS applications and shows reactivity with Human samples. The TCR2740 immunogen is human TCR Vβ3.1.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.

Specifications

Antigen TCR V beta 3.1
Applications Flow Cytometry
Classification Monoclonal
Clone 8F10
Concentration 0.2 mg/mL
Conjugate FITC
Formulation PBS with 0.5% BSA and 0.1% sodium azide
Gene TRB
Gene Accession No. P0DSE2
Gene Alias T cell receptor beta locus; TCRB; TRB; TRB@
Gene Symbols TRB
Host Species Mouse
Immunogen Human TCR V beta 3.1.
Purification Method Protein A
Quantity 200 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6957
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less