missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCP11L2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA558807

Catalog No. PIPA558807

Only null left

Description

Description

Immunogen sequence: ACLSLITNNM VGAITGGLPE LASRLTRISA VLLEGMNKET FNLKEVLNSI GIQTCVEVNK TLMERGLPTL NAEIQ Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

TCP11L2 is a protein coding gene.
Specifications

Chemical Identifiers

TCP11L2
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
TCP11L2
t-complex 11 like 2; t-complex 11, testis-specific-like 2; T-complex protein 11-like protein 2; TCP11L2
TCP11L2
Primary
Antigen affinity chromatography
0.1 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N4U5
255394
Recombinant protein corresponding to Human TCP11L2. Recombinant protein control fragment (Product #RP-97789).
Antibody

Specifications

TCP11L2
Polyclonal
Unconjugated
TCP11L2
t-complex 11 like 2; t-complex 11, testis-specific-like 2; T-complex protein 11-like protein 2; TCP11L2
Rabbit
Antigen affinity chromatography
RUO
255394
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N4U5
TCP11L2
Recombinant protein corresponding to Human TCP11L2. Recombinant protein control fragment (Product #RP-97789).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions