missing translation for 'onlineSavingsMsg'
Learn More

TCLP Extraction Fluid No. 2, Certified, LabChem™

Catalog No. p-5388762
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
Packaging:
Amber Glass
Cubitainer™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC259974 4 L Amber Glass
LC259976 10 L Cubitainer™
LC259975 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC259974 Supplier LabChem Supplier No. LC259974
Only null left

Specifications

Chemical Name or Material TCLP Extraction Fluid No. 2
Packaging Amber Glass
CAS 64-19-7, 7732-18-5
CAS Max % 99.43, 0.57
Identification Passes Test
Viscosity 1.02cSt
Density 0.999g/mL
Physical Form Liquid
Linear Formula CH3COOH
Color Colorless
pH 2.88 ±0.05
Solubility Soluble in water
Quantity 4 L
Formula Weight 60.05
Grade Certified
Show More Show Less