missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCLP Extraction Fluid No. 1, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | TCLP Extraction Fluid No. 1 |
| Packaging | Amber Glass |
| CAS Max % | 99.18, 0.57, 0.25 |
| CAS | 64-19-7, 1310-73-2 |
| Identification | Passes Test |
| Viscosity | 1.02cSt |
| Density | 0.999g/mL |
| Physical Form | Liquid |
| Linear Formula | CH3COOH |
| Color | Colorless |
| Show More |