Learn More
Environmental Express™ TCLP Bottles
With PTFE liner
$315.75 - $1,635.32
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
|
0973598
|
Environmental Express™
BC2000 1 |
2200 mL | 14 x 26 cm |
Each for $360.44
|
|
||||
|
0973269
|
Environmental Express™
BP2000 1 |
2200 mL | 11 x 32 cm |
Each for $1,635.32
|
|
||||
|
09735104
|
Environmental Express™
BF3000 4 |
3000 mL | 15 x 26 cm |
Case of 4 Each for $315.75
|
|
||||
Specifications
| Environmental Bottle | |
| PTFE |