missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520900
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520900 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520900 Supplier Invitrogen™ Supplier No. PA520900
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-20900 can be used with blocking peptide PEP-1014.

TCF3 (E2A) is a transcription factor that plays major roles in determining tissue-specific cell fate during embryogenesis, like muscle or early B-cell differentiation. Heterodimers between E2A and tissue-specific basic helix-loop-helix (bHLH) Dimers bind DNA on E-box motifs: 5'- CANNTG-3'. Binds to the kappa-E2 site in the kappa immunoglobulin gene enhancer. Deletions in E2A have been observed in a subset of pre-B-cell acute lymphoblastic leukemia (B-ALL) cases. Two alternatively spliced human isoforms have been described.

Specifications

Antigen TCF3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TCF3
Gene Accession No. P15806, P15923, P21677
Gene Alias A1; AA408400; AGM8; Alf2; AW209082; bHLHb21; class B basic helix-loop-helix protein 21; E12; E12/E47; E2A; E47; helix-loop-helix protein HE47; immunoglobulin enhancer-binding factor E12/E47; Immunoglobulin transcription factor 1; ITF1; Kappa-E2-binding factor; Me2; MGC129647; MGC129648; negative vitamin D response element-binding protein; NOL1-TCF3 fusion; Pan; Pan1; Pan2; pancreas specific transcription factor 1c; Ptf1c; TCF3; TCF-3; Tcfe2a; Transcription factor 3; transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47); transcription factor 3 variant 3; transcription factor A1; transcription factor E2a; Transcription factor E2-alpha; transcription factor ITF-1; Transcription regulator Pan; VDIR; VDR interacting repressor; vitamin D receptor-interacting repressor
Gene Symbols TCF3
Host Species Rabbit
Immunogen A 17 amino acid peptide near the amino terminus of human TCF3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171046, 21423, 6929
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less