missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCF20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150284
Description
TCF20 Polyclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a transcription factor that recognizes the platelet-derived growth factor-responsive element in the matrix metalloproteinase 3 promoter. The encoded protein is thought to be a transcriptional coactivator, enhancing the activity of transcription factors such as JUN and SP1. Mutations in this gene are associated with autism spectrum disorders. Alternative splicing results in multiple transcript variants.Chemical Identifiers
| TCF20 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9EPQ8, Q9UGU0 | |
| 21411, 366964, 6942 | |
| Recombinant Transcription factor 20 (TCF20) (Phe392-Ala524). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| TCF20 | |
| 2810438H08Rik; AR1; KIAA0292; mKIAA0292; Nuclear factor SPBP; Protein AR1; SPBP; SPRE-binding protein; stromelysin 1 PDGF responsive element binding protein; stromelysin 1 PDGF-responsive element-binding protein; stromelysin PDGF responsive element binding protein transcription factor; stromelysin-1 PDGF-responsive element-binding protein; stromelysin-1 platelet-derived growth factor-responsive element binding protein; Tcf20; TCF-20; Transcription factor 20; transcription factor 20 (AR1) | |
| TCF20 | |
| Primary | |
| Protein A |
Specifications
| TCF20 | |
| Polyclonal | |
| Unconjugated | |
| TCF20 | |
| 2810438H08Rik; AR1; KIAA0292; mKIAA0292; Nuclear factor SPBP; Protein AR1; SPBP; SPRE-binding protein; stromelysin 1 PDGF responsive element binding protein; stromelysin 1 PDGF-responsive element-binding protein; stromelysin PDGF responsive element binding protein transcription factor; stromelysin-1 PDGF-responsive element-binding protein; stromelysin-1 platelet-derived growth factor-responsive element binding protein; Tcf20; TCF-20; Transcription factor 20; transcription factor 20 (AR1) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 21411, 366964, 6942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9EPQ8, Q9UGU0 | |
| TCF20 | |
| Recombinant Transcription factor 20 (TCF20) (Phe392-Ala524). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |