missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCEA1 Recombinant Rabbit Monoclonal Antibody (23GB2330)
Description
TCEA1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | TCEA1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB2330 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TCEA1 |
| Gene Accession No. | P10711, P23193, Q4KLL0 |
| Gene Alias | GTF2S; SII; S-II; TCEA; Tcea1; Tceat; TF2S; TFIIS; transcription elongation factor A (SII) 1; transcription elongation factor A (SII), 1; transcription elongation factor A protein 1; Transcription elongation factor A protein 1-like protein; transcription elongation factor A1; transcription elongation factor S-II protein 1; Transcription elongation factor TFIIS.o; transcription factor IIS |
| Gene Symbols | TCEA1 |
| Show More |