missing translation for 'onlineSavingsMsg'
Learn More

TCEA1 Recombinant Rabbit Monoclonal Antibody (23GB2330)
GREENER_CHOICE

Catalog No. MA552502
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552502 100 μL
MA552503 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552502 Supplier Fisher Chemical Supplier No. MA552502

TCEA1 Recombinant Rabbit Monoclonal Antibody (23GB2330)

TCEA1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Specifications

Antigen TCEA1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2330
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TCEA1
Gene Accession No. P10711, P23193, Q4KLL0
Gene Alias GTF2S; SII; S-II; TCEA; Tcea1; Tceat; TF2S; TFIIS; transcription elongation factor A (SII) 1; transcription elongation factor A (SII), 1; transcription elongation factor A protein 1; Transcription elongation factor A protein 1-like protein; transcription elongation factor A1; transcription elongation factor S-II protein 1; Transcription elongation factor TFIIS.o; transcription factor IIS
Gene Symbols TCEA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TCEA1 (110-134AA).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21399, 362479, 6917
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less