missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBXA2R Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149784
Description
TBXA2R Polyclonal Antibody for Western Blot, ICC/IF, IHC
The thromboxane A2 receptor (TBXA2R) is a member of the Prostanoid Receptor subfamily. TBXA2R performs an essential role in hemostasis by interacting with thromboxane A2 to induce platelet aggregation. Thromboxane A2 has been implicated in myocardial infarct, stroke, and asthma. Additionally, thromboxane-prostanoid receptors have been implicated in the pathogenesis of cardiovascular diseases. Changes in TBXA2R function have also been linked to a hereditary bleeding disorder.Spécifications
| TBXA2R | |
| Polyclonal | |
| Unconjugated | |
| TBXA2R | |
| BDPLT13; prostanoid TP receptor; TBXA2R; Thromboxane A2 receptor; thromboxane prostanoid; Thromboxane receptor; TP; TXA2 receptor; TXA2-R; TXR2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 24816, 6915 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P21731, P34978 | |
| TBXA2R | |
| Recombinant Thromboxane Receptor (TP) (Pro3-Ser324). | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody |