missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ 20X TBS Wash Buffer for IHC/ICC

Catalog No. 5018487
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5018487 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5018487 Supplier Invitrogen™ Supplier No. 00495456

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in immunohistochemistry and immunocytochemistry protocols which require washing to remove non-specific binding. TBS Wash Buffer for IHC/ICC is recommended for eFluor™ Nanocrystal conjugated-antibodies following antibody incubation.

  • Compatible with organic dye-conjugated antibodies as well as unconjugated antibodies that may be used in two and three step staining protocols
  • The buffer is supplied as a 20x stock solution that should be diluted to 1x with distilled water

Specifications

Product Type Wash Buffer
Quantity 100 mL
For Use With (Application) Immunocytochemistry (ICC), Immunohistochemistry (IHC)
Product Line eBioscience
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.