missing translation for 'onlineSavingsMsg'
Learn More

Tazobactam sodium

Catalog No. AAJ6622603
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6622603 1 g
AAJ66226MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6622603 Supplier Thermo Scientific Chemicals Supplier No. J6622603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Beta-Lactamase inhibitor, which is used with penicillinase inhibition studies and beta-lactam antibiotics to enhance their effect. It serves as an antibacterial penicillin derivative, which inhibits the action of bacterial beta-lactamases. | CAS: 89785-84-2 | C10H12N4NaO5S | 323.279 g/mol

Tazobactam sodium is a beta-Lactamase inhibitor, which is used with penicillinase inhibition studies and beta-lactam antibiotics to enhance their effect. It serves as an antibacterial penicillin derivative, which inhibits the action of bacterial beta-lactamases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89785-84-2
Molecular Formula C10H12N4NaO5S
Molecular Weight (g/mol) 323.279
MDL Number MFCD00917472
InChI Key ZUNWFYFXHGWFRA-QVUDESDKSA-N
Synonym tazobactam sodium salt
PubChem CID 66636300
IUPAC Name (2S,3S,5R)-3-methyl-4,4,7-trioxo-3-(triazol-1-ylmethyl)-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium
SMILES CC1(C(N2C(S1(=O)=O)CC2=O)C(=O)O)CN3C=CN=N3.[Na]

Specifications

Melting Point 140°C to 147°C
Boiling Point 77°C
Flash Point >110°C (230°F)
Packaging Glass bottle
Quantity 1 g
Solubility Information Soluble in water.Soluble in water and methanol.
Formula Weight 322.27
Physical Form Powder
Chemical Name or Material Tazobactam sodium
RTECSNumber XI0191500
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only