missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Taurodeoxycholic Acid, Sodium Salt, Calbiochem™,

Catalog No. 5802215GM
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5802215GM 5 g Glass Bottle
58022150GM 50 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5802215GM Supplier MilliporeSigma™ Supplier No. 5802215GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1180-95-6
Molecular Formula C26H44NNaO7S
Molecular Weight (g/mol) 537.69
MDL Number MFCD00003671
InChI Key XDQMFTUJGWFAGC-UHFFFAOYNA-M
Synonym Sodium Taurodeoxycholate
PubChem CID 134129543
IUPAC Name sodium 4-{4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}-N-[2-(sulfonatooxy)ethyl]pentanamide
SMILES [Na+].CC(CCC(=O)NCCOS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CCC12C

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥95% (TLC)