accessibility menu, dialog, popup

UserName

CAS RN 1180-95-6

IUPAC Name: sodium 4-{4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}-N-[2-(sulfonatooxy)ethyl]pentanamide
Synonyms: sodium 4-{4,7-dihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl}-N-[2-(sulfonatooxy)ethyl]pentanamide
Molecular Weight (g/mol): 537.69
Molecular Formula: C26H44NNaO7S
InChi Key: XDQMFTUJGWFAGC-UHFFFAOYNA-M
SMILES: [Na+].CC(CCC(=O)NCCOS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CCC12C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results