missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Taurodeoxycholic Acid, Sodium Salt, Calbiochem™,
Supplier: MilliporeSigma™ 58022150GM
| Quantity | 50 g |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| 1180-95-6 | |
| 537.69 | |
| XDQMFTUJGWFAGC-UHFFFAOYNA-M | |
| 134129543 | |
| [Na+].CC(CCC(=O)NCCOS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CCC12C |
| C26H44NNaO7S | |
| MFCD00003671 | |
| Sodium Taurodeoxycholate | |
| sodium 4-{4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}-N-[2-(sulfonatooxy)ethyl]pentanamide |
Specifications
| White | |
| 50 g | |
| C26H44NNaO7S | |
| Sodium Taurodeoxycholate | |
| [Na+].CC(CCC(=O)NCCOS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CCC12C | |
| 537.69 | |
| ≥95% (TLC) |
| 1180-95-6 | |
| Glass Bottle | |
| MFCD00003671 | |
| XDQMFTUJGWFAGC-UHFFFAOYNA-M | |
| sodium 4-{4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}-N-[2-(sulfonatooxy)ethyl]pentanamide | |
| 134129543 |