missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Taurocholic Acid, Sodium Salt, ULTROL™ Grade, Calbiochem™,
$700.00 - $700.00
Chemical Identifiers
| CAS | 145-42-6 |
|---|---|
| Molecular Formula | C26H44NNaO7S |
| Molecular Weight (g/mol) | 537.688 |
| InChI Key | JAJWGJBVLPIOOH-VXFFEJGCSA-M |
| Synonym | Sodium Taurocholate |
| PubChem CID | 131632374 |
| IUPAC Name | sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate |
| SMILES | CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+] |
Chemical Identifiers
| 145-42-6 | |
| 537.688 | |
| Sodium Taurocholate | |
| sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate |
| C26H44NNaO7S | |
| JAJWGJBVLPIOOH-VXFFEJGCSA-M | |
| 131632374 | |
| CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+] |
Specifications
| White | |
| 5 g | |
| C26H44NNaO7S | |
| JAJWGJBVLPIOOH-VXFFEJGCSA-M | |
| sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate | |
| 131632374 | |
| ULTROL |
| 145-42-6 | |
| Glass Bottle | |
| Sodium Taurocholate | |
| CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+] | |
| 537.688 | |
| ≥98% (TLC) |