missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Taurocholic Acid, Sodium Salt, ULTROL™ Grade, Calbiochem™,

Catalog No. 5802185GM
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5802185GM 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5802185GM Supplier MilliporeSigma™ Supplier No. 5802185GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 145-42-6
Molecular Formula C26H44NNaO7S
Molecular Weight (g/mol) 537.688
InChI Key JAJWGJBVLPIOOH-VXFFEJGCSA-M
Synonym Sodium Taurocholate
PubChem CID 131632374
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
SMILES CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle
Percent Purity ≥98% (TLC)
Grade ULTROL