missing translation for 'onlineSavingsMsg'
Learn More

Taurocholic acid sodium salt, ≥97%, Practical grade, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210293080

 View more versions of this product

Catalog No. ICN10293080


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Taurocholic acid sodium salt
145-42-6
100 g
sodium taurocholate, taurocholic acid sodium salt, taurocholate sodium, taurocholate sodium salt, unii-m6n3th81no, monosodium n-choloyltaurinate, m6n3th81no, monosodium taurocholate, sodium n-choloyltaurinate, monosodium taurocholic acid
CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]
537.688
537.7
Practical
Brown-Yellow
Powder
C26H44NNaO7S
JAJWGJBVLPIOOH-VXFFEJGCSA-M
sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
131632374
≥97%

Chemical Identifiers

145-42-6
537.688
sodium taurocholate, taurocholic acid sodium salt, taurocholate sodium, taurocholate sodium salt, unii-m6n3th81no, monosodium n-choloyltaurinate, m6n3th81no, monosodium taurocholate, sodium n-choloyltaurinate, monosodium taurocholic acid
sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
C26H44NNaO7S
JAJWGJBVLPIOOH-VXFFEJGCSA-M
131632374
CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions