missing translation for 'onlineSavingsMsg'
Learn More

Taurine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012412
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012412 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012412 Supplier Merck Emd Millipore Supplier No. PHR11091G

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 107-35-7
Physical Form Neat
Linear Formula NH2 CH2 CH2 SO3 H
MDL Number MFCD00008197
Melting Point >300°C
Synonym 2-Aminoethanesulfonic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 125.15
Grade Certified Reference Material