missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tau Oligomeric Complex Recombinant Mouse Monoclonal Antibody (TOC1)
GREENER_CHOICE

Catalog No. MA551788
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551788 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551788 Supplier Invitrogen™ Supplier No. MA551788
Only null left

Mouse Recombinant Monoclonal Antibody

This antibody is specific for amino acids 209-224 of the Tau Oligomeric complex.

The Tau Oligomeric Complex is a protein complex found primarily in the brain. It plays a crucial role in maintaining the structure and stability of microtubules, which are important for cellular transport and neuronal function.

Specifications

Antigen Tau Oligomeric Complex
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone TOC1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias Tau Oligomer; TOC1
Host Species Mouse
Immunogen The original antibody was generated by immunizing a mouse with electro eluted cross-linked Tau dimers.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less