missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tau Oligomeric Complex Chimeric Recombinant Rabbit Monoclonal Antibody (TOC1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551567
Description
This antibody is specific for amino acids 209-224 of the Tau Oligomeric complex.
The Tau Oligomeric Complex is a protein complex found primarily in the brain. It plays a crucial role in maintaining the structure and stability of microtubules, which are important for cellular transport and neuronal function.Specifications
| Tau Oligomeric Complex Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| TOC1 | |
| Unconjugated | |
| Tau Oligomer; TOC1 | |
| The original antibody was generated by immunizing a mouse with electro eluted cross-linked Tau dimers. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |