missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAS2R16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102251
Description
Antibody detects endogenous levels of total TAS2R16.
This gene encodes a member of a family of candidate taste receptors that are members of the G protein-coupled receptor superfamily. These family members are specifically expressed by taste receptor cells of the tongue and palate epithelia. Each of these apparently intronless genes encodes a 7-transmembrane receptor protein, functioning as a bitter taste receptor. This gene is clustered with another 3 candidate taste receptor genes in chromosome 7 and is genetically linked to loci that influence bitter perception. Gustducin-coupled receptor implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in a subset of gustducin-positive taste receptor cells of the tongue. The Lys-172 polymorphism in TAS2R16 is associated with genetic susceptibility to alcoholism. Confers bitter perception of salicin to non-taster mice.Chemical Identifiers
| TAS2R16 | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| TAS2R16 | |
| candidate taste receptor T2R16; T2R16; TAS2R16; taste 2 receptor member 16; taste receptor type 2 member 16; taste receptor, type 2, member 16 | |
| TAS2R16 | |
| Primary | |
| Affinity Chromatography |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NYV7 | |
| 50833 | |
| A synthesized peptide derived from human TAS2R16(Accession Q9NYV7), corresponding to amino acid residues I142-F192. | |
| Antibody |
Specifications
| TAS2R16 | |
| Polyclonal | |
| Unconjugated | |
| TAS2R16 | |
| candidate taste receptor T2R16; T2R16; TAS2R16; taste 2 receptor member 16; taste receptor type 2 member 16; taste receptor, type 2, member 16 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 50833 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NYV7 | |
| TAS2R16 | |
| A synthesized peptide derived from human TAS2R16(Accession Q9NYV7), corresponding to amino acid residues I142-F192. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |