missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tartrate Resistant Acid Phosphatase Recombinant Rabbit Monoclonal Antibody (23GB4280)

Catalog No. MA552673
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552673 20 μL
MA552672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552673 Supplier Invitrogen™ Supplier No. MA552673
Only null left

Rabbit Recombinant Monoclonal Antibody

Tartrate Resistant Acid Phosphatase Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.

Specifications

Antigen Tartrate Resistant Acid Phosphatase
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4280
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACP5
Gene Accession No. P13686, P29288, Q05117
Gene Alias acid phosphatase 5, tartrate resistant; acid phosphatase type V; Acp5; HPAP; human purple acid phosphatase; T5ap; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; tartrate-resistant acid phosphatase type 5; TRACP; TRACP5a; TRACP5b; TRAP; TR-AP; TRAPase; TrATPase; TTRRAP; Type 5 acid phosphatase
Gene Symbols ACP5
Host Species Rabbit
Immunogen A synthesized peptide derived from human Tartrate Resistant Acid Phosphatase (150-325AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11433, 25732, 54
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less