missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tartrate Resistant Acid Phosphatase Polyclonal Antibody
Guinea Pig Polyclonal Antibody
Supplier: Invitrogen™ PA5149849
Description
Tartrate Resistant Acid Phosphatase Polyclonal Antibody for Western Blot, IHC (P)
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.Specifications
| Tartrate Resistant Acid Phosphatase | |
| Polyclonal | |
| Unconjugated | |
| ACP5 | |
| acid phosphatase 5, tartrate resistant; acid phosphatase type V; Acp5; HPAP; human purple acid phosphatase; T5ap; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; tartrate-resistant acid phosphatase type 5; TRACP; TRACP5a; TRACP5b; TRAP; TR-AP; TRAPase; TrATPase; TTRRAP; Type 5 acid phosphatase | |
| Guinea Pig | |
| Protein A | |
| RUO | |
| 100008599 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O97860 | |
| ACP5 | |
| Recombinant Acid Phosphatase 5, Tartrate Resistant (ACP5) (Tyr74-Lys321). | |
| 200 μL | |
| Primary | |
| Rabbit | |
| Antibody |