missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, Purified, Reagents

Catalog No. p-200021121
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337557 125 g
01337556 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337557 Supplier Labchem Inc Supplier No. C2116500125D2
Only null left

Tartaric Acid, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 168°C to 170°C
Boiling Point 275°C
Color Colorless/White
Grade Pure
Quantity 125 g
Packaging Glass Bottle
Physical Form Solid
Specific Gravity 1.7598
Chemical Name or Material Tartaric Acid, Purified