missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015730
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015730 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015730 Supplier Merck Emd Millipore Supplier No. PHR14721G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tartaric Acid belongs to the group of carboxylic acids, and is abundantly found in grapes and wine. It is widely used in drugs, food, and beverage industry.

Specifications

CAS 87-69-4
Ignition Point 797°F
Vapor Density 5.18 (vs. air)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula HO2 CCH(OH)CH(OH)CO2 H
MDL Number MFCD00064207
Melting Point 170°C to 172°C
Synonym L-(+)-Tartaric acid; (2 R,3 R)-(+)-Tartaric acid; L-Threaric acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 150.09
Grade Certified Reference Material
Show More Show Less