missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tartaric Acid, Granular, NF, 99.7-100.5%, Spectrum™ Chemical
TA105, 87-69-4, C4H6O6
Supplier: Spectrum Chemical Mfg Cor TA10512KG
Description
Spectrum™ Chemical Tartaric Acid, Granular, NF is used as an antioxidant, acidifier, flavor enhancer, stabilizer and sequestering agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 87-69-4 | |
| C4H6O6 | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| 2,3-dihydroxybutanedioic acid | |
| 99.7 to 100.5% | |
| 12 kg | |
| 0.005 |
| 1 | |
| MFCD00064207 | |
| OC(C(O)C(O)=O)C(O)=O | |
| 150.09 | |
| NF | |
| 0.001 | |
| Poly Pail |
Chemical Identifiers
| 87-69-4 | |
| 150.09 | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| OC(C(O)C(O)=O)C(O)=O |
| C4H6O6 | |
| MFCD00064207 | |
| 2,3-dihydroxybutanedioic acid |