missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, Granular, FCC, 99.7-100.5%, Spectrum™ Chemical

Catalog No. 18607219
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607219 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18607219 Supplier Spectrum Chemical Mfg Cor Supplier No. T10092.5KG
Only null left

T1009, 87-69-4, C4H6O6

Spectrum™ Chemical Tartaric Acid, Granular, FCC is used as an leavening additive and flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. (CAS No. 87-69-4, Molecular Formula: C4H6O6) Tartaric Acid, Granular, FCC

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

CAS Max % 1
Percent Purity 99.7 to 100.5%
Grade FCC
Quantity 2.5 kg
Ignition Residue 0.0005
Loss on Drying 0.005
Optical Rotation +12.0° to +13.0°
Packaging Poly Pail