missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. 18614313
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614313 125 g Poly Bottle
18614316 500 g Poly Bottle
18614315 2.5 kg Poly Bottle
18614314 12 kg Poly Pail
18614317 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614313 Supplier Spectrum Chemical Mfg Cor Supplier No. T1015125GM

Chemical Identifiers

CAS 87-69-4

Specifications

CAS Max % 1
Percent Purity 99%
Grade ACS
Quantity 125 g
Packaging Poly Bottle
Residue after Ignition 0.0002

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.