missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tartaric Acid, ACS Reagent, Reagents
Tartaric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1116500500A5
Specifications
| Tartaric Acid, ACS Reagent | |
| 87-69-4 | |
| Colorless/White | |
| MFCD00064207 | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| 2,3-dihydroxybutanedioic acid | |
| ACS | |
| Poly Bottle | |
| 1.7598 |
| 168°C to 170°C | |
| 275°C | |
| C4H6O6 | |
| DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid | |
| OC(C(O)C(O)=O)C(O)=O | |
| 150.09 | |
| 500 g | |
| Solid |
Chemical Identifiers
| 87-69-4 | |
| 150.09 | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| 2,3-dihydroxybutanedioic acid |
| C4H6O6 | |
| MFCD00064207 | |
| DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid | |
| OC(C(O)C(O)=O)C(O)=O |