missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, ACS Reagent, Reagents

Catalog No. 01338157
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338157 125 g
01338156 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01338157 Supplier Labchem Inc Supplier No. C1116500125D2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tartaric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym DL-Tartaric Acid, 2,3-Dihydroxysuccinic Acid, 2,3-Dihydroxybutanedioic Acid
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 168°C to 170°C
Boiling Point 275°C
Color Colorless/White
Quantity 125 g
Packaging Glass Bottle
Physical Form Solid
Specific Gravity 1.7598
Grade ACS
Chemical Name or Material Tartaric Acid, ACS Reagent