missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCT0030125
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
White Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCT0030125 125 g White Poly Wide Mouth Bottle
RDCT0030500 500 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCT0030125 Supplier Ricca Chemical Company Supplier No. RDCT0030125B1

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Percent Purity Min. 99%
Grade ACS
Quantity 125 g
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Tartaric Acid
ShelfLife 60 months