Learn More
Tartaric Acid, ACS Grade, ≥ 99.0%, LabChem™
Supplier: LabChem LC259401
Specifications
| Tartaric Acid | |
| 100 | |
| C4H6O6 | |
| MFCD00064207 | |
| Soluble in water | |
| OC(C(O)C(O)=O)C(O)=O | |
| 150.09 | |
| CHEBI:15671 | |
| ≥99.0% | |
| 500 g | |
| Poly Bottle |
| 87-69-4 | |
| White | |
| C4H6O6 | |
| l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| 2,3-dihydroxybutanedioic acid | |
| 444305 | |
| 150.09 | |
| ACS | |
| 150°C | |
| Powder |
Chemical Identifiers
| 87-69-4 | |
| 150.09 | |
| FEWJPZIEWOKRBE-UHFFFAOYNA-N | |
| 444305 | |
| 2,3-dihydroxybutanedioic acid |
| C4H6O6 | |
| MFCD00064207 | |
| l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid | |
| CHEBI:15671 | |
| OC(C(O)C(O)=O)C(O)=O |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful to aquatic life.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Danger
EINECSNumber : 201-766-0
Recommended Storage : Room Temperature