missing translation for 'onlineSavingsMsg'
Learn More

TARSL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116412
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116412 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116412 Supplier Thermo Fisher Scientific Supplier No. PA5116412
Only null left

TARSL2 Polyclonal Antibody

TARSL2 Polyclonal Antibody for Western Blot

Specifications

Antigen TARSL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TARSL2
Gene Accession No. A2RTX5, Q8BLY2
Gene Alias A530046H20Rik; probable threonine--tRNA ligase 2, cytoplasmic; Probable threonyl-tRNA synthetase 2, cytoplasmic; RGD1311308; si:zfos-508h5.3; TARS3; TARSL2; threonine--tRNA ligase; Threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase 2, cytoplasmic; probable threonine--tRNA ligase 2, cytoplasmic; threonine--tRNA ligase, cytoplasmic; threonyl-tRNA synthetase; threonyl-tRNA synthetase 3; threonyl-tRNA synthetase like 2; threonyl-tRNA synthetase-like 2; threonyl-tRNA synthetase-like protein 2; ThrRS
Gene Symbols TARSL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 380-408 amino acids from the Central region of human TARSL2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123283, 272396
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less