Learn More
PeproTech TARC Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: PeproTech 500P97BT25UG
Description
AA Sequence of recombinant protein: ARGTNVGREC CLEYFKGAIP LRKLKTWYQT SEDCSRDAIV FVTVQGRAIC SDPNNKRVKN AVKYLQSLER S. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TARC (CCL17). Anti-Human TARC (CCL17)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hTARC by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human TARC (500-P97) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hTARC. Western Blot: To detect hTARC by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hTARC is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P97BT-1MG will be provided as 2 x 500 μg
TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.Specifications
| TARC | |
| Polyclonal | |
| Biotin | |
| CCL17 | |
| A-152E5.3; ABCD-2; CC chemokine TARC; C-C motif chemokine; C-C motif chemokine 17; C-C motif chemokine ligand 17; CCL17; chemokine (C-C motif) ligand 17; SCYA17; Scya17l; small inducible cytokine subfamily A (Cys-Cys), member 17; small inducible cytokine subfamily A17; small-inducible cytokine A17; T cell-directed CC chemokine; TARC; Thymus and activation-regulated chemokine | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6361 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q92583 | |
| CCL17 | |
| E.coli-derived, 8.0 kDa Recombinant Human TARC. | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody |