Learn More
Thermo Scientific™ TaqI Buffer (10X)
Description
To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.
restriction enzymes exhibit 100% of their certified activity in the recommended buffer.
The following enzymes require unique buffers for optimal digestion:
- AarI
- AjiI
- BamHI
- BfuI
- Bpu10I
- BseXI
- Bsp143I
- Cfr9I
- Cfr10I
- Eam1105I
- Ecl136II
- Eco52I
- EcoRI
- KpnI
- Lsp1109I
- PacI
- PasI
- Ppu21I
- SacI
- ScaI
- SdaI
- SduI
- SgeI
- TaqI
Specifications
Specifications
| Concentration | 10X |
| Quantity | 1 mL |
| Product Type | TaqI Buffer |
| Research Category | Traditional Cloning |
Frequently Asked Questions (FAQs)
For optimal results with fast reaction and 100% buffer compatibility, we highly recommend using FastDigest restriction enzymes in double digestion. In certain cases however, it may be possible to perform double digestion using a mix of Thermo Scientific conventional and Fastdigest restriction enzymes. For specific recommendations, please contact our technical service with detailed information about the enzymes and DNA template you plan to use.
We recommend only 2 µl 10X Buffer in digestion of unpurified PCR products in 30 ul since salts and ions from the PCR reaction would be carried over to the digestion reaction.
Star activty may be contributed by:
• Prolonged incubation
• High enzyme concentration
• High glycerol concentration (usually 5% or higher)
• Small reaction volume
Unexpected cleavage patterns may be caused by the following reasons:
• Star activity of the restriction enzyme: Make sure to follow the reaction recommendations as specified in the protocol. Star activity may be improved by changing several key factors such as decreasing the reaction time, increasing the reaction volume, and decreasing the enzyme amount.
• Partial or incomplete cleavage (incomplete restriction reaction): Efficiency of the enzyme can be improved by adding more enzyme, prolonging the reaction time, or purifying DNA samples to remove inhibitory contaminants.
• Contamination with non-specific endonucleases: Non-specific endonucleases may be introduced to the DNA sample and/or the enzyme from improper handling, pipetting, etc.
•Improper reaction setup: Mix the digestion reaction thoroughly.
The main reason for DNA cleavage reaction failure is the presence of contaminating inhibitors in the template DNA (for example: phenol, chloroform, detergents, ethanol, excess salts, EDTA, etc.). The best way to troubleshoot is to perform control reactions:
1) negative control (experimental DNA in the reaction buffer without the restriction enzyme) to access degradation of DNA by contaminants in the DNA template and/or reaction buffer
2) positive control reaction I (digestion of highly pure control DNA with the restriction enzyme) to access reaction conditions and enzyme activity
3) positive control reaction II (highly pure control DNA + experimental DNA + Restriction Enzyme) to access possible issues with the experimental DNA.
In addition, please check for sensitivity of the restriction enzymes to template DNA methylation.
For Research Use Only. Not for use in diagnostic procedures.