missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Taq Pol Bulk (15U/μL)
Supplied with standard reaction buffer
$2,358.04
Specifications
| Concentration | 15 U/μL |
|---|---|
| For Use With (Application) | Standard PCR |
| Includes | Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
11EPTQC200
|
MP Biomedicals™
11EPTQC200 |
2000U |
Each for $2,358.04
|
|
|||||
Description
Provided with QBiogene 10X Incubation PCR Buffer which is suitable for broad range of reaction conditions.
Specifications
| 15 U/μL | |
| Standard PCR | |
| Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2 |
Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.