missing translation for 'onlineSavingsMsg'
Learn More

TAPS, 0.2M buffer soln., pH 8.5, Thermo Scientific Chemicals

$182.79 - $182.79

Chemical Identifiers

CAS 91000-53-2
Molecular Weight (g/mol) 265.256
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63268AE
View Documents
Thermo Scientific Chemicals
J63268AE
100 mL
Each for $182.79
Only null left
 
Description

Description

TAPS, is used in biochemistry and molecular biology. TAPS has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. TAPS has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TAPS, is used in biochemistry and molecular biology. TAPS has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. TAPS has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

Solubility
Soluble in water (333.33 mg/ml).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

91000-53-2
265.256

Specifications

91000-53-2
8.5
100 mL
0.2M buffer soln.
Liquid
265.256
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient Temperatures

SDS
Documents

Documents

RUO – Research Use Only