Learn More
Tantalum(V) isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol/hexane
CAS: 16761-83-4 | C15H40O5Ta | 481.428 g/mol
$264.79 - $812.04
Chemical Identifiers
| CAS | 16761-83-4 |
|---|---|
| Molecular Formula | C15H40O5Ta |
| Molecular Weight (g/mol) | 481.428 |
| MDL Number | MFCD00145347 |
| InChI Key | KWUQLGUXYUKOKE-UHFFFAOYSA-N |
| Synonym | propan-2-olate,tantalum 5+ |
| PubChem CID | 131874823 |
| IUPAC Name | propan-2-ol;tantalum |
| SMILES | CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Ta] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16761-83-4 | |
| 481.428 | |
| KWUQLGUXYUKOKE-UHFFFAOYSA-N | |
| 131874823 | |
| CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Ta] |
| C15H40O5Ta | |
| MFCD00145347 | |
| propan-2-olate,tantalum 5+ | |
| propan-2-ol;tantalum |
Specifications
| 16761-83-4 | |
| C15H40O5Ta | |
| MFCD00145347 | |
| UN1993 | |
| propan-2-olate,tantalum 5+ | |
| KWUQLGUXYUKOKE-UHFFFAOYSA-N | |
| propan-2-ol;tantalum | |
| 131874823 | |
| 99.9% | |
| Liquid | |
| (metals basis) |
| Petrol-like | |
| Ta[OCH(CH3)2]5 | |
| 25 mL | |
| Moisture sensitive | |
| Reacts with water. | |
| CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Ta] | |
| 481.428 | |
| 476.39 | |
| 10% w/v in isopropanol/hexane | |
| Tantalum(V) isopropoxide |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H304-H314-H335-H336-H361f-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only