accessibility menu, dialog, popup

UserName

CH3CH3HOCH3CH3HOCH3CH3HOCH3CH3HOCH3CH3HOTa

CAS RN 16761-83-4

IUPAC Name: pentakis(propan-2-ol) tantalum
Synonyms: pentakis(isopropyl alcohol) tantalum
Molecular Weight (g/mol): 481.43
Molecular Formula: C15H40O5Ta
InChi Key: KWUQLGUXYUKOKE-UHFFFAOYSA-N
SMILES: [Ta].CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results