missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol/hexane

Catalog No. AA40038AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40038AE 100 mL
AA40038AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40038AE Supplier Thermo Scientific Chemicals Supplier No. 040038AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16761-83-4 | C15H40O5Ta | 481.428 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16761-83-4
Molecular Formula C15H40O5Ta
Molecular Weight (g/mol) 481.428
MDL Number MFCD00145347
InChI Key KWUQLGUXYUKOKE-UHFFFAOYSA-N
Synonym propan-2-olate,tantalum 5+
PubChem CID 131874823
IUPAC Name propan-2-ol;tantalum
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Ta]

Specifications

Odor Petrol-like
Linear Formula Ta[OCH(CH3)2]5
Quantity 100 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 476.39
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 10% w/v in isopropanol/hexane
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tantalum(V) isopropoxide
Show More Show Less
Hazard Category H225-H260-H304-H314-H335-H336-H361f-H373
Hazard Statement GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only