Learn More
Tantalum(V) ethoxide, optical grade, 99.95% (metal basis)
CAS: 6074-84-6 | C10H25O5Ta | 406.25 g/mol
$63.03 - $195.95
Chemical Identifiers
| CAS | 6074-84-6 |
|---|---|
| Molecular Formula | C10H25O5Ta |
| Molecular Weight (g/mol) | 406.25 |
| MDL Number | MFCD00049785 |
| InChI Key | HSXKFDGTKKAEHL-UHFFFAOYSA-N |
| Synonym | tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide |
| PubChem CID | 160806 |
| IUPAC Name | ethanolate;tantalum(5+) |
| SMILES | CCO[Ta](OCC)(OCC)(OCC)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1603303
|
Thermo Scientific Chemicals
L1603303 |
1 g |
Each for $63.03
|
|
|||||
|
AAL1603306
|
Thermo Scientific Chemicals
L1603306 |
5 g |
Each for $195.95
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6074-84-6 | |
| 406.25 | |
| HSXKFDGTKKAEHL-UHFFFAOYSA-N | |
| 160806 | |
| CCO[Ta](OCC)(OCC)(OCC)OCC |
| C10H25O5Ta | |
| MFCD00049785 | |
| tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide | |
| ethanolate;tantalum(5+) |
Specifications
| 145°C (0.1mmHg) | |
| 6074-84-6 | |
| C10H25O5Ta | |
| MFCD00049785 | |
| 3678999 | |
| tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide | |
| CCO[Ta](OCC)(OCC)(OCC)OCC | |
| 406.25 | |
| 406.25 | |
| Optical | |
| Air and moisture sensitive | |
| Tantalum(V) ethoxide |
| 18°C to 19°C | |
| 1 g | |
| Ta(OCH2CH3)5 | |
| UN2924 | |
| 30°C (86°F) | |
| HSXKFDGTKKAEHL-UHFFFAOYSA-N | |
| ethanolate;tantalum(5+) | |
| 160806 | |
| 99.95% | |
| 1.488 | |
| 1.566 |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 228-010-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only