accessibility menu, dialog, popup

UserName

CAS RN 6074-84-6

IUPAC Name: pentaethoxytantalum
Synonyms: pentaethoxytantalum
Molecular Weight (g/mol): 406.25
Molecular Formula: C10H25O5Ta
InChi Key: HSXKFDGTKKAEHL-UHFFFAOYSA-N
SMILES: CCO[Ta](OCC)(OCC)(OCC)OCC
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (7)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)