missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) ethoxide, optical grade, 99.95% (metal basis)

Catalog No. AAL1603303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1603303 1 g
AAL1603306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1603303 Supplier Thermo Scientific Chemicals Supplier No. L1603303

CAS: 6074-84-6 | C10H25O5Ta | 406.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6074-84-6
Molecular Formula C10H25O5Ta
Molecular Weight (g/mol) 406.25
MDL Number MFCD00049785
InChI Key HSXKFDGTKKAEHL-UHFFFAOYSA-N
Synonym tantalum v ethoxide, tantalum 5+ ethanolate, tantalum ethoxide, tantalum 5+ pentaethanolate, ethanol, tantalum 5+ salt, pentaethoxytantalum v, ethanol, tantalum 5+ salt 5:1, tantalum ethylate, pentaethyl tantalate, tantalum pentaethoxide
PubChem CID 160806
IUPAC Name ethanolate;tantalum(5+)
SMILES CCO[Ta](OCC)(OCC)(OCC)OCC

Specifications

Boiling Point 145°C (0.1mmHg)
Melting Point 18°C to 19°C
Quantity 1 g
Linear Formula Ta(OCH2CH3)5
UN Number UN2924
Beilstein 3678999
Flash Point 30°C (86°F)
Formula Weight 406.25
Percent Purity 99.95%
Grade Optical
Refractive Index 1.488
Sensitivity Air and moisture sensitive
Density 1.566
Chemical Name or Material Tantalum(V) ethoxide
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 228-010-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only