Learn More
Tantalum, plasma standard solution, Specpure™ Ta 1000μg/mL, Thermo Scientific Chemicals
$149.56 - $149.56
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| MFCD00011252 |
Specifications
| Liquid | |
| MFCD00011252 | |
| 100 mL | |
| Tantalum plasma standard solution, Ta 1000μg/mL |
| TaCl5 in 2% HF | |
| UN1790 | |
| Specpure |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P390-P501c
H290-H311-H315-H319-H332
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROFLUORIC ACID
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only