missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum ICP Standard, 10,000 ppm Ta in 2% HF, Ricca Chemical
Supplier: Ricca Chemical Company PTA10KW100
Specifications
| Tantalum ICP Standard | |
| 12022-02-5,7664-39-3,7732-18-5 | |
| 10,000ppm Ta | |
| 1 mL = 10 mg Ta | |
| F7H8N2Ta | |
| Standard | |
| [NH4+].[NH4+].F[Ta-2](F)(F)(F)(F)(F)F | |
| 350.015 | |
| 71311477 |
| Natural LDPE Bottle | |
| 94.0,1.89,1.83 | |
| 98.0,1.96,1.9 | |
| Liquid | |
| Colorless | |
| RDNGDFQMYLJTJR-UHFFFAOYSA-I | |
| diazanium;heptafluorotantalum(2-) | |
| 100 mL |
Chemical Identifiers
| 12022-02-5 | |
| 350.015 | |
| 71311477 | |
| [NH4+].[NH4+].F[Ta-2](F)(F)(F)(F)(F)F |
| F7H8N2Ta | |
| RDNGDFQMYLJTJR-UHFFFAOYSA-I | |
| diazanium;heptafluorotantalum(2-) |