missing translation for 'onlineSavingsMsg'
Learn More

TAN 99.0+%, TCI America™

Catalog No. p-7118250
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50031G 1 g
A50035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50031G Supplier TCI America Supplier No. A50031G
Only null left

Chemical Identifiers

CAS 1147-56-4
Molecular Formula C13H9N3OS
Molecular Weight (g/mol) 255.30
MDL Number MFCD00021611
InChI Key GCBAKGGJQPMJLL-QINSGFPZSA-N
Synonym 1-2-thiazolylazo-2-naphthol, 1-2-thiazoylazo-2-naphthol, 2-naphthol, 1-2-thiazolylazo, 2-naphthalenol, 1-2-thiazolylazo, 1z-1-1,3-thiazol-2-ylhydrazinylidene naphthalen-2-one, 1-1,3-thiazol-2-yldiazenyl-2-naphthol, 1z-1-2-thiazolylhydrazinylidene-2-naphthalenone, 1-thiazol-2-ylazo-2-naphthol, 2-naphthalenol, 1-2-2-thiazolyl diazenyl, iomxcgdxeudzak-foclmdbbsa-n
PubChem CID 6308684
IUPAC Name (1Z)-1-[2-(1,3-thiazol-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES O=C1C=CC2=CC=CC=C2\C1=N\NC1=NC=CS1

Specifications

Melting Point 141°C
Quantity 1 g
Formula Weight 255.30
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material TAN [=1-(2-Thiazolylazo)-2-naphthol] [Spectrophotometric reagent for transition metals]