missing translation for 'onlineSavingsMsg'
Learn More

Tamsulosin Hydrochloride 98.0+%, TCI America™

Catalog No. T2749100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2749100MG 100 mg
T27491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2749100MG Supplier TCI America Supplier No. T2749100MG

Chemical Identifiers

CAS 106463-17-6
Molecular Formula C20H29ClN2O5S
Molecular Weight (g/mol) 444.971
MDL Number MFCD00922997
InChI Key ZZIZZTHXZRDOFM-XFULWGLBSA-N
Synonym tamsulosin hydrochloride, omnic, tamsulosin hcl, pradif, flomax, urolosin, secotex, josir, alna, omic
PubChem CID 5362376
ChEBI CHEBI:9399
IUPAC Name 5-[(2R)-2-[2-(2-ethoxyphenoxy)ethylamino]propyl]-2-methoxybenzenesulfonamide;hydrochloride
SMILES CCOC1=CC=CC=C1OCCNC(C)CC2=CC(=C(C=C2)OC)S(=O)(=O)N.Cl

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 100 mg
Formula Weight 444.97
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tamsulosin Hydrochloride
RTECSNumber DB2430000
TSCA No