missing translation for 'onlineSavingsMsg'
Learn More

Tamsulosin hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012396
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012396 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012396 Supplier Merck Emd Millipore Supplier No. PHR1524200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tamsulosin hydrochloride is a subtypeselective a1A and a1D adrenoceptor antagonist, which exists in two enantiomeric forms, of which the R-isomer is the pharmaceutically active component. It is used to reduce urinary obstruction and is also involved in relieving the symptoms associated with symptomatic benign prostatic hyperplasia.

Specifications

Packaging Ampule
CAS 106463-17-6
Linear Formula C20H28N2O5 S · HCl
MDL Number MFCD00922997
Synonym Tamsulosin hydrochloride; 5-[(2 R)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide hydrochloride
Type Certified Reference Material
Quantity 200 mg
Formula Weight 444.97
Grade Certified Reference Material