missing translation for 'onlineSavingsMsg'
Learn More

Tamsulosin hydrochloride, 98+%

Catalog No. AAJ61999MC
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61999MC 100 mg
AAJ61999MA 10 mg
AAJ61999MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61999MC Supplier Thermo Scientific Chemicals Supplier No. J61999MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An alpha-1 adrenoceptor antagonist | CAS: 106463-17-6 | C20H29ClN2O5S | 444.971 g/mol

An alpha-1 adrenoceptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106463-17-6
Molecular Formula C20H29ClN2O5S
Molecular Weight (g/mol) 444.971
MDL Number MFCD00922997
InChI Key ZZIZZTHXZRDOFM-XFULWGLBSA-N
Synonym tamsulosin hydrochloride, omnic, tamsulosin hcl, pradif, flomax, urolosin, secotex, josir, alna, omic
PubChem CID 5362376
ChEBI CHEBI:9399
IUPAC Name 5-[(2R)-2-[2-(2-ethoxyphenoxy)ethylamino]propyl]-2-methoxybenzenesulfonamide;hydrochloride
SMILES CCOC1=CC=CC=C1OCCNC(C)CC2=CC(=C(C=C2)OC)S(=O)(=O)N.Cl

Specifications

Melting Point 228°C to 230°C
Color White
Quantity 100 mg
Merck Index 14,9049
Formula Weight 444.97
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Tamsulosin hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber DB2430000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only